siRNA Details
| Gene Symbol: Cpa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013083.2 | NP_001013101.2 | 1 | 23 | ||
| GenBank mRNA | CF249075.1 | - | 1 | 23 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpa2| Gene Symbol: Cpa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013083.2 | NP_001013101.2 | 1 | 23 | ||
| GenBank mRNA | CF249075.1 | - | 1 | 23 | ||