siRNA Details
| Gene Symbol: Prkar2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019264.2 | NP_062137.1 | 1, 2 | 372 | ||
| GenBank mRNA | AF533978.1 | - | 1, 2 | 376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkar2a| Gene Symbol: Prkar2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019264.2 | NP_062137.1 | 1, 2 | 372 | ||
| GenBank mRNA | AF533978.1 | - | 1, 2 | 376 | ||