siRNA Details
| Gene Symbol: Pcbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007601.2 | NP_001007602.1 | 2 | 137 | ||
| XM_006256414.1 | XP_006256476.1 | 3 | 395 | |||
| GenBank mRNA | FM060466.1 | - | 2 | 137 | ||
| FM067129.1 | - | 2 | 114 | |||
| L04537.1 | - | 1 | 40 | |||
| M83740.1 | - | 2 | 333 | |||

Pcbd1
Genomic Map