siRNA Details
Gene Symbol: Pcbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007601.2 | NP_001007602.1 | 4 | 337 | ||
XM_006256414.1 | XP_006256476.1 | 5 | 595 | |||
GenBank mRNA | DV724713.1 | - | 2 | 146 | ||
FM060466.1 | - | 4 | 337 | |||
FM067129.1 | - | 4 | 314 | |||
M83740.1 | - | 4 | 533 |
