siRNA Details
| Gene Symbol: Rarres2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013427.1 | NP_001013445.1 | 3 | 323 | ||
| XM_006236415.2 | XP_006236477.1 | 3 | 350 | |||
| XM_006236416.1 | XP_006236478.1 | 3 | 411 | |||
| GenBank mRNA | BC091177.1 | - | 3 | 323 | 7323591 | |
| FQ209660.1 | - | 3 | 338 | |||
| FQ209771.1 | - | 3 | 337 | |||
| FQ210577.1 | - | 3 | 338 | |||
| FQ210699.1 | - | 3 | 338 | |||
| FQ228347.1 | - | 3 | 372 | |||

Rarres2
Genomic Map