siRNA Details
| Gene Symbol: Gabrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183327.1 | NP_899156.1 | 1 | 39 | ||
| XM_006246125.2 | XP_006246187.1 | 1 | 602 | |||
| GenBank mRNA | AY574252.1 | - | 1 | 39 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrg2| Gene Symbol: Gabrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183327.1 | NP_899156.1 | 1 | 39 | ||
| XM_006246125.2 | XP_006246187.1 | 1 | 602 | |||
| GenBank mRNA | AY574252.1 | - | 1 | 39 | ||