siRNA Details
Gene Symbol: Kcnj5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017297.2 | NP_058993.1 | 2 | 192 | ||
GenBank mRNA | BC087022.1 | - | 2 | 192 | 7192366 | |
D50135.1 | - | 2 | 145 | |||
L35771.1 | - | 2 | 197 | |||
X83584.1 | - | 1 | 16 |
