siRNA Details
Gene Symbol: Cacna1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031601.3 | NP_113789.2 | 3 | 938 | ||
XM_008767985.1 | XP_008766207.1 | 3 | 956 | |||
XM_008767986.1 | XP_008766208.1 | 3 | 956 | |||
XM_008767987.1 | XP_008766209.1 | 3 | 956 | |||
XM_008767988.1 | XP_008766210.1 | 3 | 956 | |||
XM_008767989.1 | XP_008766211.1 | 3 | 956 | |||
XM_008767990.1 | XP_008766212.1 | 3 | 956 | |||
XM_008767991.1 | XP_008766213.1 | 3 | 956 | |||
XM_008767992.1 | XP_008766214.1 | 3 | 956 | |||
XM_008767993.1 | XP_008766215.1 | 3 | 956 | |||
XM_008767994.1 | XP_008766216.1 | 3 | 956 | |||
XM_008767995.1 | XP_008766217.1 | 3 | 956 | |||
XM_008767996.1 | XP_008766218.1 | 3 | 956 | |||
XM_008767997.1 | XP_008766219.1 | 3 | 956 | |||
XM_008767998.1 | XP_008766220.1 | 3 | 956 | |||
XM_008767999.1 | XP_008766221.1 | 3 | 956 | |||
XM_008768000.1 | XP_008766222.1 | 3 | 956 | |||
XM_008768001.1 | XP_008766223.1 | 3 | 956 | |||
GenBank mRNA | AF027984.1 | - | 3 | 938 | ||
AF125161.1 | - | 3 | 613 | |||
AF290212.2 | - | 3 | 569 | |||
EF116282.1 | - | 3 | 406 |
