siRNA Details
| Gene Symbol: Hivep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024137.1 | NP_077051.1 | 3 | 1616 | ||
| GenBank mRNA | D37951.1 | - | 3 | 1616 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hivep2| Gene Symbol: Hivep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024137.1 | NP_077051.1 | 3 | 1616 | ||
| GenBank mRNA | D37951.1 | - | 3 | 1616 | ||