siRNA Details
| Gene Symbol: Mrpl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106596.1 | NP_001100066.1 | 2 | 262 | ||
| XM_006236635.1 | XP_006236697.1 | 2 | 219 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl35| Gene Symbol: Mrpl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106596.1 | NP_001100066.1 | 2 | 262 | ||
| XM_006236635.1 | XP_006236697.1 | 2 | 219 | |||