siRNA Details
| Gene Symbol: Rnf181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007647.1 | NP_001007648.1 | 2 | 463 | ||
| GenBank mRNA | BC079313.1 | - | 2 | 463 | 7134895 | |
| FQ213488.1 | - | 1 | 112 | |||
| FQ230795.1 | - | 1 | 62 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf181| Gene Symbol: Rnf181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007647.1 | NP_001007648.1 | 2 | 463 | ||
| GenBank mRNA | BC079313.1 | - | 2 | 463 | 7134895 | |
| FQ213488.1 | - | 1 | 112 | |||
| FQ230795.1 | - | 1 | 62 | |||