siRNA Details
| Gene Symbol: Capg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013086.1 | NP_001013104.1 | 3, 4 | 499 | ||
| XM_006236661.2 | XP_006236723.1 | 4, 5 | 339 | |||
| XM_006236662.2 | XP_006236724.1 | 4, 5 | 432 | |||
| XM_006236663.2 | XP_006236725.1 | 4, 5 | 566 | |||
| XM_006236666.2 | XP_006236728.1 | 3, 4 | 286 | |||
| GenBank mRNA | BC079104.1 | - | 3, 4 | 499 | 7125038 | |

Capg
Genomic Map