siRNA Details
Gene Symbol: Htra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106599.2 | NP_001100069.1 | 1, 2 | 525 | ||
GenBank mRNA | BC087588.1 | - | 1, 2 | 505 | ||
BC158760.1 | - | 1, 2 | 525 | 7103492 |
