siRNA Details
| Gene Symbol: Bola3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106601.1 | NP_001100071.1 | 2 | 179 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bola3| Gene Symbol: Bola3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106601.1 | NP_001100071.1 | 2 | 179 | ||