siRNA Details
Gene Symbol: Tex261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017537.1 | NP_001017537.1 | 5 | 407 | ||
XM_008763059.1 | XP_008761281.1 | 5 | 485 | |||
XM_008763060.1 | XP_008761282.1 | 5 | 613 | |||
XM_008763061.1 | XP_008761283.1 | 5 | 360 | |||
GenBank mRNA | BC091302.1 | - | 5 | 407 | 7309682 | |
FQ214220.1 | - | 5 | 419 | |||
FQ218231.1 | - | 5 | 418 |
