siRNA Details
| Gene Symbol: Gclm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017305.2 | NP_059001.1 | 5 | 519 | ||
| GenBank mRNA | BC078867.1 | - | 5 | 519 | 7110139 | |
| L22191.1 | - | 5 | 457 | |||
| S65555.1 | - | 5 | 457 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gclm| Gene Symbol: Gclm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017305.2 | NP_059001.1 | 5 | 519 | ||
| GenBank mRNA | BC078867.1 | - | 5 | 519 | 7110139 | |
| L22191.1 | - | 5 | 457 | |||
| S65555.1 | - | 5 | 457 | |||