siRNA Details
Gene Symbol: Cct7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106603.1 | NP_001100073.1 | 9 | 1645 | ||
GenBank mRNA | FQ214319.1 | - | 9 | 1015 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cct7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106603.1 | NP_001100073.1 | 9 | 1645 | ||
GenBank mRNA | FQ214319.1 | - | 9 | 1015 |