siRNA Details
Gene Symbol: Nfu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106606.2 | NP_001100076.2 | 7 | 659 | ||
XM_008763063.1 | XP_008761285.1 | 7 | 892 | |||
GenBank mRNA | FM064033.1 | - | 6 | 485 | ||
FM092780.1 | - | 1 | 42 |
