siRNA Details
| Gene Symbol: Nfu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106606.2 | NP_001100076.2 | 8 | 729 | ||
| XM_008763063.1 | XP_008761285.1 | 8 | 962 | |||
| GenBank mRNA | FM064033.1 | - | 7 | 555 | ||
| FM092780.1 | - | 2 | 112 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfu1| Gene Symbol: Nfu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106606.2 | NP_001100076.2 | 8 | 729 | ||
| XM_008763063.1 | XP_008761285.1 | 8 | 962 | |||
| GenBank mRNA | FM064033.1 | - | 7 | 555 | ||
| FM092780.1 | - | 2 | 112 | |||