siRNA Details
| Gene Symbol: Gkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039686.1 | NP_001034775.1 | 3 | 127 | ||
| XM_008763064.1 | XP_008761286.1 | 2 | 1655 | |||
| XM_008763065.1 | XP_008761287.1 | 6 | 611 | |||
| GenBank mRNA | AY943907.1 | - | 3 | 127 | ||

Gkn2
Genomic Map