siRNA Details
| Gene Symbol: Pik3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022185.2 | NP_071521.2 | 2 | 454 | ||
| XM_008771096.1 | XP_008769318.1 | 1 | 1614 | |||
| GenBank mRNA | BC089805.1 | - | 2 | 454 | 7381613 | |
| D64046.1 | - | 1 | 14 | |||

Pik3r2
Genomic Map