siRNA Details
| Gene Symbol: Copg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031822.3 | NP_001026992.1 | 16 | 1631 | ||
| XM_008763066.1 | XP_008761288.1 | 16 | 1836 | |||
| GenBank mRNA | AB158425.1 | - | 16 | 1631 | ||
| AB158426.1 | - | 16 | 1631 | |||
| BC129074.1 | - | 16 | 1722 | 7107382 | ||

Copg1
Genomic Map