siRNA Details
| Gene Symbol: Hdac11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106610.2 | NP_001100080.2 | 10 | 938 | ||
| GenBank mRNA | BC166430.1 | - | 10 | 938 | 7131565 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdac11| Gene Symbol: Hdac11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106610.2 | NP_001100080.2 | 10 | 938 | ||
| GenBank mRNA | BC166430.1 | - | 10 | 938 | 7131565 | |