siRNA Details
| Gene Symbol: Mrps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025408.1 | NP_001020579.1 | 3 | 309 | ||
| GenBank mRNA | BC097381.1 | - | 3 | 309 | 7445826 | |
| FQ210488.1 | - | 3 | 292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps25| Gene Symbol: Mrps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025408.1 | NP_001020579.1 | 3 | 309 | ||
| GenBank mRNA | BC097381.1 | - | 3 | 309 | 7445826 | |
| FQ210488.1 | - | 3 | 292 | |||