siRNA Details
| Gene Symbol: Eif4e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106612.1 | NP_001100082.1 | 5 | 399 | ||
| XM_008763159.1 | XP_008761381.1 | 5 | 813 | |||
| XM_008763160.1 | XP_008761382.1 | 7 | 883 | |||
| XM_008763161.1 | XP_008761383.1 | 6 | 747 | |||
| GenBank mRNA | BC168936.1 | - | 5 | 386 | 7124999 | |

Eif4e3
Genomic Map