siRNA Details
| Gene Symbol: Shq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134713.1 | NP_001128185.1 | 1313 | |||
| XM_008763162.1 | XP_008761384.1 | 1321 | ||||
| GenBank mRNA | BC168172.1 | - | 1313 | 9026449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shq1| Gene Symbol: Shq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134713.1 | NP_001128185.1 | 1313 | |||
| XM_008763162.1 | XP_008761384.1 | 1321 | ||||
| GenBank mRNA | BC168172.1 | - | 1313 | 9026449 | ||