siRNA Details
| Gene Symbol: Crbn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015003.2 | NP_001015003.1 | 4 | 467 | ||
| GenBank mRNA | AY973953.1 | - | 4 | 454 | ||
| BC099779.1 | - | 4 | 452 | |||
| BC127455.1 | - | 4 | 459 | 7112865 | ||
| CB567139.1 | - | 4 | 477 | |||

Crbn
Genomic Map