siRNA Details
| Gene Symbol: Rad52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106617.2 | NP_001100087.2 | 11 | 1043 | ||
| XM_008763232.1 | XP_008761454.1 | 11 | 1774 | |||
| XM_008763233.1 | XP_008761455.1 | 11 | 1779 | |||
| XM_008763234.1 | XP_008761456.1 | 11 | 1772 | |||
| XM_008763235.1 | XP_008761457.1 | 11 | 1774 | |||
| XM_008763236.1 | XP_008761458.1 | 11 | 1773 | |||
| XM_008763237.1 | XP_008761459.1 | 11 | 1774 | |||
| XM_008763238.1 | XP_008761460.1 | 10 | 1693 | |||

Rad52
Genomic Map