siRNA Details
| Gene Symbol: Usp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106619.1 | NP_001100089.1 | 4 | 415 | ||
| XM_006237338.1 | XP_006237400.1 | 4 | 408 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp5| Gene Symbol: Usp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106619.1 | NP_001100089.1 | 4 | 415 | ||
| XM_006237338.1 | XP_006237400.1 | 4 | 408 | |||