siRNA Details
| Gene Symbol: Ddx47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015005.2 | NP_001015005.2 | 11, 12 | 1252 | ||
| XM_006237518.1 | XP_006237580.1 | 12, 13 | 1309 | |||
| GenBank mRNA | BC091696.1 | - | 11, 12 | 1245 | 7301732 | |

Ddx47
Genomic Map