siRNA Details
Gene Symbol: LOC297756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013928.2 | NP_001013950.2 | 2 | 597 | ||
GenBank mRNA | AY383664.1 | - | 2 | 597 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC297756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013928.2 | NP_001013950.2 | 2 | 597 | ||
GenBank mRNA | AY383664.1 | - | 2 | 597 |