siRNA Details
| Gene Symbol: Cntnap5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047866.1 | NP_001041331.1 | 518 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntnap5c| Gene Symbol: Cntnap5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047866.1 | NP_001041331.1 | 518 | |||