siRNA Details
| Gene Symbol: Zfp362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191612.1 | NP_001178541.1 | 6 | 937 | ||
| XM_006238922.1 | XP_006238984.1 | 6 | 1026 | |||
| XM_006238923.2 | XP_006238985.1 | 7 | 1521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp362| Gene Symbol: Zfp362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191612.1 | NP_001178541.1 | 6 | 937 | ||
| XM_006238922.1 | XP_006238984.1 | 6 | 1026 | |||
| XM_006238923.2 | XP_006238985.1 | 7 | 1521 | |||