siRNA Details
| Gene Symbol: Gem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106637.1 | NP_001100107.1 | 510 | |||
| GenBank mRNA | BC168988.1 | - | 460 | 8369626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gem| Gene Symbol: Gem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106637.1 | NP_001100107.1 | 510 | |||
| GenBank mRNA | BC168988.1 | - | 460 | 8369626 | ||