siRNA Details
| Gene Symbol: Smim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106644.1 | NP_001100114.1 | 4 | 758 | ||
| NM_001201373.1 | NP_001188302.1 | 2 | 234 | |||
| NM_001201374.1 | NP_001188303.1 | 3 | 380 | |||
| NM_001201375.1 | NP_001188304.1 | 3 | 624 | |||
| XM_006237987.2 | XP_006238049.1 | 4 | 663 | |||
| XM_006237988.2 | XP_006238050.1 | 3 | 546 | |||
| GenBank mRNA | BC167112.1 | - | 2 | 234 | 6887495 | |
| DY544415.1 | - | 3 | 380 | |||
| FQ226097.1 | - | 3 | 585 | |||

Smim8
Genomic Map