siRNA Details
| Gene Symbol: Pole3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007652.2 | NP_001007653.1 | 4 | 432 | ||
| GenBank mRNA | BC081988.1 | - | 4 | 432 | 7129121 | |
| BC083800.1 | - | 4 | 432 | 7108773 | ||
| Gene Symbol: LOC100362333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729829.4 | XP_002729875.1 | 1 | 493 | ||

Pole3
Genomic Map