siRNA Details
| Gene Symbol: Bnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106666.1 | NP_001100136.1 | 7 | 3010 | ||
| XM_008763775.1 | XP_008761997.1 | 7 | 4285 | |||
| XM_008763776.1 | XP_008761998.1 | 6 | 4201 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bnc2| Gene Symbol: Bnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106666.1 | NP_001100136.1 | 7 | 3010 | ||
| XM_008763775.1 | XP_008761997.1 | 7 | 4285 | |||
| XM_008763776.1 | XP_008761998.1 | 6 | 4201 | |||