siRNA Details
| Gene Symbol: Bnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106666.1 | NP_001100136.1 | 6 | 1008 | ||
| XM_008763775.1 | XP_008761997.1 | 6 | 2283 | |||
| XM_008763776.1 | XP_008761998.1 | 5 | 2199 | |||
| XM_008763777.1 | XP_008761999.1 | 6 | 2282 | |||

Bnc2
Genomic Map