siRNA Details
| Gene Symbol: Mtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047867.1 | NP_001041332.1 | 8 | 801 | ||
| XM_006238382.2 | XP_006238444.1 | 6 | 712 | |||
| GenBank mRNA | AY325240.1 | - | 8 | 801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtap| Gene Symbol: Mtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047867.1 | NP_001041332.1 | 8 | 801 | ||
| XM_006238382.2 | XP_006238444.1 | 6 | 712 | |||
| GenBank mRNA | AY325240.1 | - | 8 | 801 | ||