siRNA Details
| Gene Symbol: Fggy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013932.2 | NP_001013954.2 | 3 | 324 | ||
| XM_006238410.2 | XP_006238472.1 | 3 | 962 | |||
| XM_006238413.2 | XP_006238475.1 | 4 | 535 | |||
| XM_008763856.1 | XP_008762078.1 | 3 | 324 | |||
| GenBank mRNA | BC090077.1 | - | 2 | 149 | 7107754 | |
| CB316436.1 | - | 3 | 327 | |||

Fggy
Genomic Map