siRNA Details
| Gene Symbol: Gpx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106673.1 | NP_001100143.1 | 2 | 431 | ||
| GenBank mRNA | FQ229888.1 | - | 2 | 376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpx7| Gene Symbol: Gpx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106673.1 | NP_001100143.1 | 2 | 431 | ||
| GenBank mRNA | FQ229888.1 | - | 2 | 376 | ||