siRNA Details
| Gene Symbol: Magoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100536.1 | NP_001094006.1 | 1 | 119 | ||
| XM_006238510.2 | XP_006238572.1 | 1 | 166 | |||
| GenBank mRNA | BQ196337.1 | - | 5 | 548 | ||
| CK845348.1 | - | 5 | 514 | |||
| DQ359101.1 | - | 1 | 48 | |||

Magoh
Genomic Map