siRNA Details
| Gene Symbol: Creb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013092.2 | NP_001013110.1 | 2 | 469 | ||
| XM_006238050.1 | XP_006238112.1 | 2 | 495 | |||
| XM_008763672.1 | XP_008761894.1 | 3 | 299 | |||
| GenBank mRNA | BC062241.1 | - | 2 | 472 | 6887761 | |
| FQ219084.1 | - | 2 | 447 | |||
| KJ817427.1 | - | 2 | 313 | |||

Creb3
Genomic Map