siRNA Details
| Gene Symbol: Nasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005543.1 | NP_001005543.1 | 7 | 1484 | ||
| XM_006238646.2 | XP_006238708.1 | 7 | 1430 | |||
| XM_008763993.1 | XP_008762215.1 | 7 | 1507 | |||
| GenBank mRNA | BC081913.1 | - | 7 | 1484 | 7112076 | |
| FQ212041.1 | - | 6 | 1424 | |||

Nasp
Genomic Map