siRNA Details
| Gene Symbol: Dmap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015006.1 | NP_001015006.1 | 4, 5 | 496 | ||
| GenBank mRNA | BC092651.1 | - | 4, 5 | 496 | 7319534 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmap1| Gene Symbol: Dmap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015006.1 | NP_001015006.1 | 4, 5 | 496 | ||
| GenBank mRNA | BC092651.1 | - | 4, 5 | 496 | 7319534 | |