siRNA Details
| Gene Symbol: Ppie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047868.1 | NP_001041333.1 | 5, 6 | 245 | ||
| XM_006238796.2 | XP_006238858.1 | 4, 5 | 260 | |||
| XM_006238797.2 | XP_006238859.1 | 4, 5 | 269 | |||
| GenBank mRNA | AY325152.1 | - | 5, 6 | 245 | ||
| BC158560.1 | - | 4, 5 | 210 | 9102303 | ||

Ppie
Genomic Map