siRNA Details
| Gene Symbol: Ppie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047868.1 | NP_001041333.1 | 10 | 771 | ||
| GenBank mRNA | AY325152.1 | - | 10 | 771 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppie| Gene Symbol: Ppie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047868.1 | NP_001041333.1 | 10 | 771 | ||
| GenBank mRNA | AY325152.1 | - | 10 | 771 | ||