siRNA Details
| Gene Symbol: Tekt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011977.1 | NP_001011977.1 | 11 | 1284 | ||
| XM_006238860.1 | XP_006238922.1 | 11 | 1456 | |||
| XM_006238861.2 | XP_006238923.1 | 10 | 1217 | |||
| GenBank mRNA | BC078990.1 | - | 11 | 1284 | 7112575 | |

Tekt2
Genomic Map