siRNA Details
| Gene Symbol: Dhdds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011978.1 | NP_001011978.1 | 7 | 694 | ||
| XM_006239014.2 | XP_006239076.1 | 7 | 809 | |||
| GenBank mRNA | BC085875.1 | - | 7 | 694 | 7191470 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhdds| Gene Symbol: Dhdds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011978.1 | NP_001011978.1 | 7 | 694 | ||
| XM_006239014.2 | XP_006239076.1 | 7 | 809 | |||
| GenBank mRNA | BC085875.1 | - | 7 | 694 | 7191470 | |