siRNA Details
| Gene Symbol: Zfp593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106689.1 | NP_001100159.1 | 1 | 157 | ||
| GenBank mRNA | BC166512.1 | - | 1 | 113 | 7128768 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp593| Gene Symbol: Zfp593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106689.1 | NP_001100159.1 | 1 | 157 | ||
| GenBank mRNA | BC166512.1 | - | 1 | 113 | 7128768 | |